missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCF4 Antibody (TCF4/1705), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08236AF532
This item is not returnable.
View return policy
Description
TCF4 Monoclonal antibody specifically detects TCF4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin), Protein Array
Specifications
| TCF4 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| BHLHB19, bHLHb19E2-2, Class B basic helix-loop-helix protein 19, Immunoglobulin transcription factor 2, ITF-2, ITF2PTHS, MGC149724, SEF-2, SEF2-1, SEF2-1A, SEF2-1B, SEF2MGC149723, SL3-3 enhancer factor 2, TCF-4, transcription factor 4 | |
| Recombinant human TCF4 protein fragment (around aa 365-671) (exact sequence is proprietary) (Uniprot: P15884 ) | |
| 100 μg | |
| Transcription Factors and Regulators | |
| 6925 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Western Blot, ELISA, Immunohistochemistry (Paraffin), Protein Array | |
| TCF4/1705 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction