missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TCF7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578191
This item is not returnable.
View return policy
Description
Positive Control: HepG2, Jurkat Predicted Reactivity: Mouse (97%), Bovine (95%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TCF7 (transcriptional factor 7) is a transcriptional activator involved in T-cell lymphocyte differentiation. Necessary for the survival of CD4+ and CD8+ immature thymocytes. Isoforms for TCF7 lacking the N-terminal CTNNB1 binding domain cannot fulfill this role. TCF7 binds to the T-lymphocyte-specific enhancer element (5'-WWCAAAG-3') found in the promoter of the CD3E gene. It may also act as feedback transcriptional repressor of CTNNB1 and TCF7L2 target genes. TLE1, TLE2, TLE3, and TLE4 repress transactivation mediated by TCF7 and CTNNB1.
Specifications
| TCF7 | |
| Polyclonal | |
| Unconjugated | |
| TCF7 | |
| AI465550; HMG-box; OTTHUMP00000159390; OTTHUMP00000159391; T cell factor-1; T-cell factor 1; T-cell specific; T-cell-specific transcription factor 1; Tcf1; TCF-1; Tcf7; TCF-7; Transcription factor 7; transcription factor 7 (T-cell specific, HMG-box); transcription factor 7, T cell specific; transcription factor 7, T-cell specific | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 21414, 363595, 6932 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| P36402, Q00417 | |
| TCF7 | |
| Recombinant protein encompassing a sequence within the center region of human TCF7. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction