missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TCF7L1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15574082
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15574082 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15574082 Supplier Invitrogen™ Supplier No. PA540327

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: NDELIPFQDE GGEEQEPSSD SASAQRDLDE VKSSLVNESE NQSSSSDSEA Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rat: 100%.

This gene encodes a member of the T cell factor/lymphoid enhancer factor family of transcription factors. These transcription factors are activated by beta catenin, mediate the Wnt signaling pathway and are antagonized by the transforming growth factor beta signaling pathway. The encoded protein contains a high mobility group-box DNA binding domain and participates in the regulation of cell cycle genes and cellular senescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCF7L1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TCF7L1
Gene Accession No. Q9HCS4
Gene Alias bHLHb21; HMG box transcription factor 3; T-cell factor 3; Tcf3; TCF-3; TCF7L1; transcription factor 3; transcription factor 7 like 1; transcription factor 7 like 1 (T cell specific, HMG box); transcription factor 7-like 1; transcription factor 7-like 1 (T-cell specific, HMG box); transcription factor 7-like 1 (T-cell specific, HMG-box); transcription factor 7-like 2 (T-cell specific, HMG box)
Gene Symbols TCF7L1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human TCF7L1 (aa 35-84).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83439
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.