missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCHP Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25931-1-AP-150UL
This item is not returnable.
View return policy
Description
TCHP may act as a ′capping′ or ′branching′ protein for keratin filaments in the cell periphery. This protein may regulate K8/K18 filament and desmosome organization mainly at the apical or peripheral regions of simple epithelial cells.Specifications
| TCHP | |
| Polyclonal | |
| Unconjugated | |
| TCHP | |
| Protein TCHP, TCHP, Tumor suppressor protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 77832, 84260 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.41 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q3TVW5, Q9BT92 | |
| Tchp | |
| TCHP Fusion Protein Ag23199 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction