missing translation for 'onlineSavingsMsg'
Learn More

TCIRG1 Antibody [Alexa Fluor« 405], Novus Biologicals Biologicals™

Product Code. 30503767 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30503767 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30503767 Supplier Novus Biologicals Supplier No. NBP335541AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TCIRG1 Polyclonal antibody specifically detects TCIRG1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCIRG1
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias a3, Atp6i, ATP6N1Cspecific 116-kDa vacuolar proton pump subunit, ATP6V0A3T-cell immune response cDNA 7, OC-116, OC-116 kDa, OC-116kDa, OC116Vph1, OPTB1, Osteoclastic proton pump 116 kDa subunit, Stv1, T-cell immune regulator 1, T-cell immune response cDNA7 protein, T-cell, immune regulator 1, T-cell, immune regulator 1, ATPase, H+ transporting, lysosomal V0 protein a, T-cell, immune regulator 1, ATPase, H+ transporting, lysosomal V0 protein A3, T-cell, immune regulator 1, ATPase, H+ transporting, lysosomal V0 protein aisoform 3, T-cell, immune regulator 1, ATPase, H+ transporting, lysosomal V0 subunit A3, TIRC7ATPase, H+ transporting, 116kD, vacuolar proton translocating ATPase 116 kDa subunit A, Vacuolar proton translocating ATPase 116 kDa subunit a isoform 3, V-ATPase 116 kDa isoform a3, V-ATPase 116-kDa, V-type proton ATPase 116 kDa subunit a, V-type proton ATPase 116 kDa subunit a isoform 3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-130 of human TCIRG1 (NP_006010.2).,, Sequence:, MGSMFRSEEVALVQLFLPTAAAYTCVSRLGELGLVEFRDLNASVSAFQRRFVVDVRRCEELEKTFTFLQEEVRRAGLVLPPPKGRLPAPPPRDLLRIQEETERLAQELRDVRGNQQALRAQLHQLQLHAA
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 10312
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.