Learn More
TCL1 (T-Cell Marker) Mouse, Clone: TCL1/2078, enQuireBio™
Mouse Monoclonal Antibody
€ 199.00 - € 478.00
Specifications
| Antigen | TCL1 (T-Cell Marker) |
|---|---|
| Clone | TCL1/2078 |
| Concentration | 200μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15911128
|
EnquireBio
8115-MSM1-P0 |
20 μg | |||||||
|
15921128
|
EnquireBio
8115-MSM1-P1 |
100 μg |
€ 554.40 € 478.00 / 100µg Save € 76.40 13% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| TCL1 (T-Cell Marker) | |
| 200μg/mL | |
| Monoclonal | |
| Mouse | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Recombinant human TCL1 protein fragment (apprx. aa 2-109) | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
| 14kDa |
| TCL1/2078 | |
| Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| 10232 | |
| IgG1 κ | |
| Protein A/G purified from Bioreactor concentrate. | |
| Nuclear and Cytoplasmic | |
| Ramos or Raji cells. Tonsil or T-cell Lymphoma. |