missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCL1A Antibody (TCL1/2747R), HRP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08384H
This item is not returnable.
View return policy
Description
TCL1A Monoclonal antibody specifically detects TCL1A in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| TCL1A | |
| Monoclonal | |
| HRP | |
| Oncogene TCL1, Oncogene TCL-1, Protein p14 TCL1, T-cell leukemia/lymphoma 1A, T-cell lymphoma-1, T-cell lymphoma-1A, TCL1T-cell leukemia/lymphoma protein 1A | |
| Recombinant human TCL1A protein fragment (around aa 2-109) (exact sequence is proprietary) (Uniprot: P56279) | |
| 100 μg | |
| Cancer | |
| 8115 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| TCL1/2747R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction