missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCLP Standard 1: Cd, Se @ 5; Ag, As, Cr, Pb @ 25; Ba @ 500 μg/mL in 2% HNO3, VHG™
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Description
TCLP Standard 1: Cd, Se @ 5; Ag, As, Cr, Pb @ 25; Ba @ 500 μg/mL in 2% HNO3
Specifications
Specifications
| Chemical Name or Material | Cadmium/Selenium/Arsenic/Chromium/Lead/Silver/Barium |
| CAS | 7440-43-9, 7782-49-2, 7440-38-2, 7440-47-3, 7439-92-1, 7440-22-4 |
| Methods | ICP/ICP-MS |
| Analyte or Component Names | Cadmium/Selenium/Arsenic/Chromium/Lead/Silver/Barium |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Dilute HNO3 |
| Quantity | 100 mL |
| Concentration or Composition (by Analyte or Components) | Cadmium 5 μg/mL/Selenium 5 μg/mL/Arsenic 25 μg/mL/Chromium 25 μg/mL/Lead 25 μg/mL/Silver 25 μg/mL/Barium 500 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction