missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TCN2 Polyclonal Antibody

Description
Reconstitute at 0.2 mg/mL in sterile PBS.
This gene encodes a member of the vitamin B12-binding protein family. This family of proteins, alternatively referred to as R binders, is expressed in various tissues and secretions. This plasma protein binds cobalamin and mediates the transport of cobalamin into cells. This protein and other mammalian cobalamin-binding proteins, such as transcobalamin I and gastric intrisic factor, may have evolved by duplication of a common ancestral gene. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
| Antigen | TCN2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with trehalose and no preservative |
| Gene | TCN2 |
| Gene Accession No. | P20062 |
| Gene Alias | AW208754; D22S676; D22S750; II; macrocytic anemia; TC; TC II; TC TC D22S676; Tc2; TC-2; TCII; Tcn2; Tcn-2; Tcn2p; transcobalamin; transcobalamin 2; transcobalamin II; transcobalamin II precursor; transcobalamin II; macrocytic anemia; transcobalamin-2; vitamin B12-binding protein 2 |
| Gene Symbols | TCN2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?