Learn More
Invitrogen™ TCN2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120946
Description
Positive test controls include: SW480, MCF7, A-549, U-251MG, Mouse kidney, Mouse lung. The target is usually found in the following locations: Secreted. Immunogen sequence: EMCEIPEMDS HLVEKLGQHL LPWMDRLSLE HLNPSIYVGL RLSSLQAGTK EDLYLHSLKL GYQQCLLGSA FSEDDGDCQG KPSMGQLALY LLALRANWHD HKGHPHTSYY QYGLGILALC LHQKRVHDSV VDKLLYAVEP FHQGHHSVDT AAMAGLAFTC LKRSNFNPGR RQRITMAIRT VREEILKAQT PEGHFGNVYS TPLALQFLMT SPMRGAELGT ACLKARVALL AS.
This gene encodes a member of the vitamin B12-binding protein family. This family of proteins, alternatively referred to as R binders, is expressed in various tissues and secretions. This plasma protein binds cobalamin and mediates the transport of cobalamin into cells. This protein and other mammalian cobalamin-binding proteins, such as transcobalamin I and gastric intrisic factor, may have evolved by duplication of a common ancestral gene. Alternative splicing results in multiple transcript variants.
Specifications
| TCN2 | |
| Polyclonal | |
| Unconjugated | |
| TCN2 | |
| AW208754; D22S676; D22S750; II; macrocytic anemia; TC; TC II; TC TC D22S676; Tc2; TC-2; TCII; Tcn2; Tcn-2; Tcn2p; transcobalamin; transcobalamin 2; transcobalamin II; transcobalamin II precursor; transcobalamin II; macrocytic anemia; transcobalamin-2; vitamin B12-binding protein 2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 21452, 6948 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O88968, P20062 | |
| TCN2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 19-250 of human TCN2 (NP_001171655.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |