missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TCP-1 gamma Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106304
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CCT3.
The protein encoded by this gene is a molecular chaperone that is a member of the chaperonin containing TCP1 complex (CCT), also known as the TCP1 ring complex (TRiC). This complex consists of two identical stacked rings, each containing eight different proteins. Unfolded polypeptides enter the central cavity of the complex and are folded in an ATP-dependent manner. The complex folds various proteins, including actin and tubulin. Alternate transcriptional splice variants have been characterized for this gene. In addition, a pseudogene of this gene has been found on chromosome 8.
Specifications
| TCP-1 gamma | |
| Polyclonal | |
| Unconjugated | |
| Cct3 | |
| AL024092; CCT3; CCTG; CCT-gamma; chaperonin containing TCP1 subunit 3; chaperonin containing Tcp1, subunit 3 (gamma); chaperonin subunit 3 (gamma); hTRiC5; Matricin; mTRiC-P5; OTTHUMP00000025735; PIG48; T-complex protein 1 subunit gamma; T-complex protein 1, gamma subunit; TCP1 (t-complex-1) ring complex, polypeptide 5; TCP-1-gamma; Tcp1-rs3; TRIC5; TriC-P5 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12462, 295230, 7203 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P49368, P80318, Q6P502 | |
| Cct3 | |
| A synthesized peptide derived from human CCT3(Accession P49368), corresponding to amino acid residues K353-L403. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction