missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCP1 alpha Antibody (23c (CTA-123)), DyLight 755, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-50328IR
This item is not returnable.
View return policy
Description
TCP1 alpha Monoclonal specifically detects TCP1 alpha in Mouse, Rat, Canine, Chicken, Hamster samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| TCP1 alpha | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TCP1 | |
| C-terminal half of the full length murine TCP protein | |
| 0.1 mL | |
| Cancer, Hypoxia, Membrane Trafficking and Chaperones, Stem Cell Markers | |
| 6950 | |
| Mouse, Rat, Canine, Chicken, Hamster | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 23c (CTA-123) | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| CCT1T-complex protein 1 subunit alpha, CCTa, CCT-alpha, D6S230E, tailless complex polypeptide 1, t-complex 1, T-complex protein 1, alpha subunit, TCP-1-alpha | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Does not recognize primate, xenopus or yeast TCP-1. | |
| Store at 4C in the dark. | |
| IgG2c |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction