missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCP1 alpha Antibody (23c (CTA-123)), Janelia Fluor™ 646, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-50328JF646
This item is not returnable.
View return policy
Description
TCP1 alpha Monoclonal specifically detects TCP1 alpha in Mouse, Rat, Canine, Chicken, Hamster samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| TCP1alpha | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TCP1 | |
| C-terminal half of the full length murine TCP protein | |
| 0.1 mL | |
| Primary | |
| Does not recognize primate, xenopus or yeast TCP-1. | |
| Store at 4°C in the dark. | |
| IgG2c |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 23c (CTA-123) | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| CCT1T-complex protein 1 subunit alpha, CCTa, CCT-alpha, D6S230E, tailless complex polypeptide 1, t-complex 1, T-complex protein 1, alpha subunit, TCP-1-alpha | |
| Rat | |
| Protein G purified | |
| Cancer, Hypoxia, Membrane Trafficking and Chaperones, Stem Cell Markers | |
| 6950 | |
| Canine, Chicken, Hamster, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction