missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ TCP1-beta Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29102
This item is not returnable.
View return policy
Description
TCP1-beta Polyclonal antibody specifically detects TCP1-beta in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| TCP1-beta | |
| Polyclonal | |
| Tris-buffered Saline 0.1% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Cctb, CCT-beta, CCTBPRO1633,99D8.1, chaperonin containing t-complex polypeptide 1, beta subunit, chaperonin containing t-complex polypeptide 1, subunit 2, chaperonin containing TCP1, subunit 2 (beta), MGC142074, MGC142076, T-complex protein 1 subunit beta, T-complex protein 1, beta subunit, TCP-1-beta | |
| between 484 and 534 | |
| 20 μg | |
| Cell Cycle and Replication, Membrane Trafficking and Chaperones | |
| 10576 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction