Learn More
TCP1-delta Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-41361AF647
Product Code. 18785360
  • € 633.00 / 0.10mL
Description
TCP1-delta Polyclonal antibody specifically detects TCP1-delta in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TCP1-delta
Polyclonal
Western Blot
Western Blot
Alexa Fluor 647
Store at 4°C in the dark.