missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCP1-delta Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-32385
This item is not returnable.
View return policy
Description
TCP1-delta Polyclonal specifically detects TCP1-delta in Human samples. It is validated for Western Blot.
Specifications
| TCP1-delta | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| CCT4 | |
| Recombinant protein encompassing a sequence within the center region of human TCP-1 delta. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Tested in human. Predicted cross-reactivity based on sequence identity: Xenopus laevis (92%) and Xenopus tropicalis (92%). Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.59mg/mL | |
| Western Blot 1:500-1:3000 | |
| CCTD, CCT-DELTA, chaperonin containing t-complex polypeptide 1, delta subunit, chaperonin containing TCP1, subunit 4 (delta), MGC126165, SRBMGC126164, Stimulator of TAR RNA-binding, T-complex protein 1 subunit delta, TCP-1-delta | |
| Rabbit | |
| 58 kDa | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 10575 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction