missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCP10 Antibody (OTI2C2) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74474
This item is not returnable.
View return policy
Description
TCP10 Monoclonal specifically detects TCP10 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| TCP10 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:2000, Immunohistochemistry 1:150 | |
| MGC34049, t-complex 10 (a murine tcp homolog), t-complex 10 (mouse), t-complex 10 homolog (mouse), T-complex protein 10A homolog, TCP10A | |
| Mouse | |
| 35.4 kDa | |
| 100 μg | |
| 6953 | |
| Human | |
| Lyophilized |
| Western Blot, Immunohistochemistry | |
| OTI2C2 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| TCP10 | |
| Full length human recombinant protein of human TCP10 (NP_004601) produced in 293T cell. | |
| Affinity Purified | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction