missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCP10 Monoclonal Antibody (OTI2C2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809068S
This item is not returnable.
View return policy
Description
TCP10 Monoclonal antibody specifically detects TCP10 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| TCP10 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P30405 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10105 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2C2 | |
| Unconjugated | |
| Ppif | |
| t-complex 10; t-complex 10 (a murine tcp homolog); t-complex 10 homolog; T-complex protein 10A homolog; TCP10; TCP10A | |
| Full length human recombinant protein of human PPIF produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction