missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCR alpha Antibody (34), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52715B
This item is not returnable.
View return policy
Description
TCR alpha Monoclonal antibody specifically detects TCR alpha in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF
Specifications
| TCR alpha | |
| Monoclonal | |
| Biotin | |
| C), D, FLJ22602, J, MGC117436, MGC22624, MGC23964, MGC71411, T cell receptor alpha locus, T-cell antigen receptor, alpha polypeptide, T-cell receptor, alpha (V, TCRA, TCRD, TRA | |
| Purified recombinant soluble TCR from the T-cell hybridoma 85.33, corresponding to V alpha 11 | |
| 0.1 mL | |
| Adaptive Immunity, Cell Biology, Immunology, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
| 6955 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, ELISA, CyTOF | |
| 34 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction