missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCR alpha Antibody (34), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52715PECY55
This item is not returnable.
View return policy
Description
TCR alpha Monoclonal antibody specifically detects TCR alpha in Human samples. It is validated for Flow Cytometry
Specifications
| TCR alpha | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| C), D, FLJ22602, J, MGC117436, MGC22624, MGC23964, MGC71411, T cell receptor alpha locus, T-cell antigen receptor, alpha polypeptide, T-cell receptor, alpha (V, TCRA, TCRD, TRA | |
| Purified recombinant soluble TCR from the T-cell hybridoma 85.33, corresponding to V alpha 11 | |
| 0.1 mL | |
| Adaptive Immunity, Cell Biology, Immunology, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
| 6955 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| Flow Cytometry | |
| 34 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction