missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TCR alpha/beta Monoclonal Antibody (IP26), NovaFluor™ Yellow 660, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
€ 178.00 - € 392.00
Specifications
| Antigen | TCR alpha/beta |
|---|---|
| Clone | IP26 |
| Concentration | 0.2 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30055412
|
Invitrogen™
H086T03Y04-A |
100 Tests |
€ 392.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30054828
|
Invitrogen™
H086T02Y04-A |
25 Tests |
€ 178.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TCR alpha/beta Monoclonal antibody specifically detects TCR alpha/beta in Human samples. It is validated for Flow CytometrySpecifications
| TCR alpha/beta | |
| 0.2 μg/Test | |
| Monoclonal | |
| Mouse | |
| Human | |
| P01848, P04435, P0DSE1, P0DSE2 | |
| 28589, 28755, 6955, 6957 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| IP26 | |
| Flow Cytometry | |
| NovaFluor Yellow 660 | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| FLJ22602; IMD7; LOC290071; MGC117436; MGC22624; MGC23964; MGC71411; RATTCB; RATTCBC1; RGD1359684; similar to RIKEN cDNA A430107P09 gene; T cell receptor beta locus; T3/TCR complex; TCB; TCBC1; t-cell antigen receptor; T-cell receptor alpha constant; T-cell receptor beta chain; T-cell receptor V alpha; tcr alpha; TCR alpha/ beta; TCR beta; TCRA; Tcrb; TRA; Tra29; TRAC; TRB; TRB@; TRCA | |
| IgG1 κ | |
| Affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title