Learn More
Invitrogen™ TCR alpha/beta Recombinant Armenian Hamster Monoclonal Antibody (H57-597)

Description
mAb H57-597 reacts with surface receptors on all alpha-beta TCR bearing cells, but does not react with receptors on gamma-delta T cells. H57-597 is directed to an epitope within the FG loop of the C region of TCR beta chain. Preparation: Hamsters were immunised with affinity purified DO-11.10 TCR, and produced detectable anti-TCR antibodies which were judged by the ability of the anti-sera to immunoprecipitate TCR from surface labelled DO-11.10 T cell hybridomas. Hamster 57 was selected for hybridisation, and B cell hybridomas were produced using polyethylene glycol fusion of hamster splenic B cells to the mouse myeloma variant. Selected mAb were screened after subsequent culturing for their ability to immunoprecipitate TCR from detergent lysates of surface-radiolabelled murine T cell hybridomas.
Specifications
Specifications
| Antigen | TCR alpha/beta |
| Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation |
| Classification | Recombinant Monoclonal |
| Clone | H57-597 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% ProClin 300 |
| Gene | TCRA |
| Gene Accession No. | P01849 |
| Gene Alias | FLJ22602; IMD7; LOC290071; MGC117436; MGC22624; MGC23964; MGC71411; RATTCB; RATTCBC1; RGD1359684; similar to RIKEN cDNA A430107P09 gene; T cell receptor beta locus; T3/TCR complex; TCB; TCBC1; t-cell antigen receptor; T-cell receptor alpha constant; T-cell receptor beta chain; T-cell receptor V alpha; tcr alpha; TCR alpha/ beta; TCR beta; TCRA; Tcrb; TRA; Tra29; TRAC; TRB; TRB@; TRCA |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.