missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCR beta Antibody (H57-597) [Janelia Fluor™ 635], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-20118JF635
This item is not returnable.
View return policy
Description
TCR beta Monoclonal antibody specifically detects TCR beta in Mouse samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunoprecipitation,Immunocytochemistry/ Immunofluorescence
Specifications
| TCR beta | |
| Monoclonal | |
| Janelia Fluor 635 | |
| T cell receptor beta chain, TCRB, TCRbeta, Tib | |
| Hamsters were immunised with affinity purified DO-11.10 TCR, and produced detectable anti-TCR antibodies which were judged by the ability of the anti-sera to immunoprecipitate TCR from surface labelled DO-11.10 T cell hybridomas. Hamster 57 was selected for hybridisation, and B cell hybridomas were produced using polyethylene glycol fusion of hamster splenic B cells to the mouse myeloma variant. Selected mAb were screened after subsequent culturing for their ability to immunoprecipitate TCR from detergent lysates of surface-radiolabelled murine T cell hybridomas. | |
| 0.1 mL | |
| Adaptive Immunity, Cell Biology, Diabetes Research, Immunology, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
| 21577 | |
| Store at 4°C in the dark. | |
| IgG λ |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry/Immunofluorescence | |
| H57-597 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction