Learn More
Invitrogen™ TCR beta Monoclonal Antibody (H57-597), Brilliant Ultra Violet™ 496, eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
€ 197.00 - € 399.00
Specifications
| Antigen | TCR beta |
|---|---|
| Clone | H57-597 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18416598
![]() |
Invitrogen™
364-5961-82 |
100 μg |
€ 399.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18436448
|
Invitrogen™
364-5961-80 |
25 μg |
€ 197.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivning
TCR beta Monoclonal antibody specifically detects TCR beta in Mouse samples. It is validated for Flow CytometrySpecifikationer
| TCR beta | |
| 0.2 mg/mL | |
| Monoclonal | |
| Armenian Hamster | |
| Mouse | |
| bTCR; FLJ22602; MGC117436; MGC22624; MGC23964; MGC71411; T cell receptor beta chain; t-cell antigen receptor; T-cell receptor; TCR beta; Tcra; Tcrb; TCRbeta; TCRD; Tib; TRA | |
| IgG | |
| Affinity chromatography | |
| Tcrb |
| H57-597 | |
| Flow Cytometry | |
| Brilliant Ultraviolet 496 | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| 21577 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel