Learn More
TCR β Chain Hamster anti-Mouse, APC, Clone: H57-597, BD
Hamster Monoclonal Antibody
€ 61.00 - € 244.00
Specifications
| Clone | H57-597 |
|---|---|
| Concentration | 0.2mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Host Species | Hamster |
| Product Code | Brand | Quantity | Conjugate | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Conjugate | Price | Quantity & Availability | ||||
|
15870749
|
BD Biosciences
561080 |
25 μg | APC |
€ 61.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
|
15847058
|
BD Biosciences
553174 |
0.1 mg | APC |
€ 244.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
|
15860379
|
BD Biosciences
560656 |
50 μg | APC-Cy7 |
€ 121.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The H57-597 antibody reacts with a common epitope of the β chain of the T-cell Receptor (TCR) complex on αβ TCR-expressing thymocytes and peripheral T lymphocytes and NK1.1+ thymocytes and NK-T cells of all mouse strains tested. It does not react with γδ TCR-bearing T cells. In the fetal and adult thymus, the TCR β-chain may form homodimers or pair with the pre-TCR α-chain on the surface of immature thymocytes before expression of the TCR α-chain. Plate-bound or soluble H57-597 antibody activates αβ TCR-bearing T cells, and plate-bound mAb can induce apoptotic death.
Specifications
| H57-597 | |
| Flow Cytometry | |
| Hamster | |
| Mouse | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| 0.2mg/mL | |
| Monoclonal | |
| RUO | |
| TCR affinity-purified from mouse T-cell hybridoma DO-11.10 | |
| Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.