missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCR gamma/delta Antibody (B1.1), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-43121MFV450
This item is not returnable.
View return policy
Description
TCR gamma/delta Monoclonal antibody specifically detects TCR gamma/delta in Human samples. It is validated for Flow Cytometry
Specifications
| TCR gamma/delta | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| B1.1 | |
| Flow Cytometry | |
| T cell receptor gamma locus, T-cell antigen receptor, gamma polypeptide, T-cell rearranging gene, gamma, T-cell receptor, gamma cluster, TCRG, TRG | |
| The immunogen for this antibody was gamma delta TCR. | |
| 0.1 mL | |
| Adaptive Immunity, Immunology, Stem Cells | |
| 6965 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction