missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCR gamma/delta Antibody (GL-3) [Janelia Fluor™ 669], Novus Biologicals™
Hamster Monoclonal Antibody
Brand: Novus Biologicals FAB7297JF669
This item is not returnable.
View return policy
Description
TCR gamma/delta Monoclonal antibody specifically detects TCR gamma/delta in Mouse samples. It is validated for Flow Cytometry,CyTOF-reported
Specifications
| TCR gamma/delta | |
| Monoclonal | |
| Janelia Fluor 669 | |
| T cell receptor gamma locus, T-cell antigen receptor, gamma polypeptide, T-cell rearranging gene, gamma, T-cell receptor, gamma cluster, TCRG, TRG | |
| Mouse intraepithelial lymphocytes | |
| RUO | |
| Primary | |
| Mouse | |
| Liquid |
| Flow Cytometry, CyTOF | |
| GL-3 | |
| 50mM Sodium Borate | |
| Hamster | |
| 0.1 mL | |
| Adaptive Immunity, Immunology, Stem Cells | |
| 6965 | |
| Store at 4°C in the dark. | |
| IgG2 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction