missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCR V beta 10 Antibody (G101), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64281V
This item is not returnable.
View return policy
Description
TCR V beta 10 Monoclonal specifically detects TCR V beta 10 in Rat samples. It is validated for Flow Cytometry.
Specifications
| TCR V beta 10 | |
| Monoclonal | |
| DyLight 405 | |
| T cell receptor beta locus, T-cell antigen receptor, beta polypeptide, T-cell receptor, beta cluster, T-cell receptor, beta cluster, TCRB, TRB | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 6957 | |
| Rat | |
| Purified |
| Flow Cytometry | |
| G101 | |
| Flow Cytometry | |
| TRB | |
| T cell clone expressing the TCR V beta 10 gene product | |
| 0.125 mL | |
| Primary | |
| NB100-64281 is specific for the T-cell receptor V beta 10 gene product. Approximately 4.2% of unseparated LEW lymph node cells were detected by this clone. Reactivity is restricted to alpha/beta T cells, of which 6.7% labelled (1). | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction