missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCR V beta 3 Antibody (JOVI.3), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50356APC
This item is not returnable.
View return policy
Description
TCR V beta 3 Monoclonal antibody specifically detects TCR V beta 3 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Functional Assay, CyTOF
Specifications
| TCR V beta 3 | |
| Monoclonal | |
| APC | |
| P09693 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Functional Assay, CyTOF | |
| JOVI.3 | |
| PBS | |
| T cell receptor beta locus, T-cell antigen receptor, beta polypeptide, T-cell receptor, beta cluster, T-cell receptor, beta cluster, TCRB, TRB | |
| Thymus, spleen and mesenteric lymph nodes isolated from a mouse transgenic for human Vb3 TcR. | |
| 0.1 mL | |
| Adaptive Immunity, Cell Biology, Diabetes Research, Immunology, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
| 6957 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction