missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCR V beta 5b Antibody (W112), Novus Biologicals™
Mouse Monoclonal Antibody
€ 200.00
Specifications
| Antigen | TCR V beta 5b |
|---|---|
| Clone | W112 |
| Concentration | 0.15 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Flow Cytometry 1 - 2 μg, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Functional |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Functional Assay |
Description
TCR V beta 5b Monoclonal specifically detects TCR V beta 5b in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
| TCR V beta 5b | |
| 0.15 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Functional Assay | |
| Unconjugated | |
| Mouse | |
| Human | |
| T cell receptor beta locus, T-cell antigen receptor, beta polypeptide, T-cell receptor, beta cluster, T-cell receptor, beta cluster, TCRB, TRB | |
| TCB | |
| IgG1 | |
| Protein G purified | |
| This antibody reacts with Human. |
| W112 | |
| Western Blot 1:100 - 1:2000, Flow Cytometry 1 - 2 μg, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Functional | |
| Monoclonal | |
| Ascites | |
| RUO | |
| P04435 | |
| 6957 | |
| Human TCR Vb5.3 | |
| Primary | |
| Store at 4°C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title