Learn More
Invitrogen™ TCR V beta 8b Monoclonal Antibody (MX-6)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ TCR1750
Product Code. 11898831
  • € 502.00 / 200µg
Description
TCR1750 targets TCR V beta 8b in FACS, IHC(F), IP, and TCA applications and shows reactivity with Human samples. The TCR1750 immunogen is human TCR Vbeta8.
The ability of T cell receptors (TCR) to discriminate foreign from self-peptides presented by major histocompatibility complex (MHC) class II molecules is essential for an effective adaptive immune response. TCR recognition of self-peptides has been linked to autoimmune disease. Mutant self-peptides have been associated with tumors. Engagement of TCRs by a family of bacterial toxins know as superantigens has been responsible for toxic shock syndrome. Autoantibodies to V beta segments of T cell receptors have been isolated from patients with rheumatoid arthritis (RA) and systemic lupus erythematosus (SLE). The autoantibodies block TH1-mediated inflammatory autodestructive reactions and are believed to be a method by which the immune system compensates for disease (ref5). T Cell and TCR Diversity Most human T cells express the TCR alpha-beta and either CD4 or CD8 molecule (single positive, SP). A small number of T cells lack both CD4 and CD8 (double negative, DN). Increased percentages of alpha-beta DN T cells have been identified in some autoimmune and immunodeficiency disorders. Gamma-delta T cells are primarily found within the epithelium. They show less TCR diversity and recognize antigens differently than alpha-beta T cells. Subsets of gamma-delta T cells have shown antitumor and immunoregulatory activity.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TCR V beta 8b
Monoclonal
0.15 mg/mL
PBS with 0.5% BSA and 0.1% sodium azide
P0DSE2
TRB
Human TCR Vbeta8.
200 μg
Primary
Human
Antibody
IgG2a
Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, T-Cell Activation
MX-6
Unconjugated
TRB
T cell receptor beta locus; TCRB; TRB; TRB@
Mouse
Protein G
RUO
6957
-20°C