Learn More
Invitrogen™ TCR V beta F1 Monoclonal Antibody (8A3), APC, eBioscience™

Mouse Monoclonal Antibody
€ 371.00
Specifications
| Antigen | TCR V beta F1 |
|---|---|
| Clone | 8A3 |
| Concentration | 5 μL/Test |
| Content And Storage | 4°C, store in dark, DO NOT FREEZE! |
| Applications | Flow Cytometry |
Description
Description: This 8A3 monoclonal antibody recognizes human TCR beta F1, a common framework determinant on the beta-subunit of the T cell receptor. Expression of TCR beta F1 has been described as a potential diagnostic marker indicating a favorable outcome for tumor patients with not otherwise specified Nodal Peripheral T-cell Lymphoma (PTCL-NOS). Furthermore, a lack of surface and cytoplasmic TCR beta F1 was also found in small cell variant of T-cell prolymphocytic Leukemia. This antibody will not stain the native form of the TCR and will require fixing and permeabilizing cells. Applications Reported: This 8A3 antibody has been reported for use in flow cytometric analysis, immunofluorescent staining of frozen tissue sections, and immunocytochemistry. Applications Tested: This 8A3 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.
The T Cell Receptor alpha beta (TCRab) is a heterodimeric membrane receptor that consists of the highly variable alpha and beta chains expressed as part of a complex with the invariant CD3 proteins. TCRab participates in the activation of T-cells in response to an antigen presented in the context of MHC. Engagement of the TCRab initiates signaling cascades that ultimately result in cellular proliferation, differentiation, cytokine production, and/or activation-induced cell death. These signaling cascades regulate T-cell development, homeostasis, activation, acquisition of effector functions and apoptosis. Cells expressing TCRab constitute the vast majority of T cells in both human and mouse.Specifications
| TCR V beta F1 | |
| 5 μL/Test | |
| Flow Cytometry | |
| APC | |
| Mouse | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| P0DSE2 | |
| 6957 | |
| Human TCR beta chain constant region. | |
| Antibody |
| 8A3 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Monoclonal | |
| Liquid | |
| IgG1 κ | |
| Human | |
| TRB | |
| T cell receptor beta locus; tcr beta f1; TCRB; TRB; TRB@ | |
| TRB | |
| Primary | |
| Affinity chromatography |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.