missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCR V delta 1 Antibody (TS8.2), Novus Biologicals™
Mouse Monoclonal Antibody
€ 213.00 - € 482.00
Specifications
| Antigen | TCR V delta 1 |
|---|---|
| Clone | TS8.2 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 1 - 2 ug, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500 |
| Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18430452
|
Novus Biologicals
NBP2-22506-50ug |
50 ug |
€ 213.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18110012
|
Novus Biologicals
NBP2-22506 |
200 μg |
€ 482.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TCR V delta 1 Monoclonal specifically detects TCR V delta 1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
| TCR V delta 1 | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Mouse | |
| Human | |
| 6964 | |
| Human TCR V delta 1 | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| TS8.2 | |
| Flow Cytometry 1 - 2 ug, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| T cell receptor delta locus, T-cell antigen receptor, delta polypeptide, T-cell receptor, delta (V,D,J,C), TCRD, TCRD, TRD@, TCRDV1, TRD@ | |
| TRD | |
| IgG1 | |
| Protein G purified | |
| This antibody reacts with Human. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title