Learn More
Invitrogen™ TCR V delta 1 Monoclonal Antibody (TS8.2), PE-Cyanine7, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen™ 25-5679-42
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Mouse Monoclonal Antibody
The T Cell Receptor gamma-delta (TCR gamma delta) is a heterodimeric membrane receptor that consists of the highly variable gamma and delta chains expressed as part of a complex with the invariant CD3 proteins. Gamma-delta T cells are less frequent than alpha-beta T cells but compose a substantial part of T cells found at epithelial borders including the gut mucosa resident intraepithelial lymphocytes (IEL). The antigenic stimuli required for gamma-delta T cell receptor activation are not well understood and are thought to be mainly MHC class-independent.
Spécification
| TCR V delta 1 | |
| Monoclonal | |
| 5 μL/Test | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| 0 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 6964 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid | |
| Responsibly packaged |
| Flow Cytometry | |
| TS8.2 | |
| PE-Cyanine7 | |
| TRD | |
| TRD | |
| Full length protein of Human TCR delta v1. | |
| 100 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |