missing translation for 'onlineSavingsMsg'
Learn More

TCS JNK 5a, MedChemExpress

Product Code. p-200101600
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16422957

Brand: Medchem Express HY15881

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TCS JNK 5a is a potent JNK3 inhibitor with a pIC50 of 6.7. TCS JNK 5a also inhibits JNK2 with a pIC50 of 6.5.

TCS JNK 5a is a potent JNK3 inhibitor with a pIC50 of 6.7. TCS JNK 5a also inhibits JNK2 with a pIC50 of 6.5.

Specifications

Chemical Name or Material TCS JNK 5a
Physical Form Solid
CAS 312917-14-9
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C20H16N2OS
For Use With (Application) Cancer-Kinase/protease
Synonym JNK Inhibitor IX
Solubility Information DMSO : ≥ 51 mg/mL (153.42 mM)
SMILES O=C(C1=C2C=CC=CC2=CC=C1)NC3=C(C#N)C(CCCC4)=C4S3
Molecular Weight (g/mol) 332.42
Formula Weight 332.42
Percent Purity 98.96%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.