missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ TCS OX2 29
GREENER_CHOICE

Product Code. 18013802
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18013802 10 mg 10mg
18015672 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18013802 Supplier Tocris Bioscience™ Supplier No. 3371/10
  • ONLINE EXCLUSIVE PRICE
    € 322.00
    € 304.50 / 10mg
    Save € 17.50
    5% Off
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Potent and selective OX2 antagonist

Potent and selective OX2 receptor antagonist (IC50 = 40 nM). Displays >250-fold selectivity for OX2 over OX1 and a range of receptors, ion channels and transporters. Inhibits orexin A induced IP3 accumulation and ERK1/2 phosphorylation in CHO cells transfected with the OX2 receptor.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1610882-30-8
Molecular Formula C23H32ClN3O3

Specifications

Quantity 10 mg
Formula Weight 433.97
Purity >98%
Molecular Weight (g/mol) 433.977
Chemical Name or Material TCS OX2 29
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.