missing translation for 'onlineSavingsMsg'
Learn More

TCS PIM-1 1, MedChemExpress

Product Code. 16477458
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16477458

Brand: Medchem Express HY18086

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TCS PIM-1 1 (SC 204330) is a potent, selective and ATP-competitive Pim-1 kianse inhibitor with an IC50 of 50 nM, displays good selectivity over Pim-2 and MEK1/MEK2 (IC50s >20000 nM).

TCS PIM-1 1 (SC 204330) is a potent, selective and ATP-competitive Pim-1 kianse inhibitor with an IC50 of 50 nM, displays good selectivity over Pim-2 and MEK1/MEK2 (IC50s >20000 nM).

Specifications

Chemical Name or Material TCS PIM-1 1
CAS 491871-58-0
Color Yellow
Purity Grade Notes Research
Molecular Formula C18H11BrN2O2
Quantity 10 mg
Synonym SC 204330
Solubility Information DMSO : 100 mg/mL (272.33 mM; Need ultrasonic)
SMILES OC1=C(C(N2)=CC(C3=CC=CC=C3)=C(C#N)C2=O)C=C(Br)C=C1
Molecular Weight (g/mol) 367.2
Formula Weight 367.2
Percent Purity 98.01%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.