missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCTE1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 188.00 - € 470.00
Specifications
| Antigen | TCTE1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18617731
|
Novus Biologicals
NBP2-94592-0.02ml |
0.02 mL |
€ 188.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18686891
|
Novus Biologicals
NBP2-94592-0.1ml |
0.1 mL |
€ 470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TCTE1 Polyclonal antibody specifically detects TCTE1 in Human, Mouse, Rat samples. It is validated for Western BlotSpécification
| TCTE1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology | |
| PBS (pH 7.3), 50% glycerol | |
| 202500 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| D6S46, FLJ50041, MGC33600, T-complex-associated testis-expressed protein 1, t-complex-associated-testis-expressed 1, tcte-1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human TCTE1 (NP_872345.2). MQDTVTTSALLDPSHSSVSTQDNSSTGGHTSSTSPQLSKPSITPVPAKSRNPHPRANIRRMRRIIAEDPEWSLAIVPLLTELCIQHIIRNFQKNPILKQMLPEHQQKVLNHLSPDLPLAVTANLIDSENYWLRCCMHRWPVCHVAHHGGSWKRMFFERHLENLLKHFIPGTTDPAVILDLLPLCRNYVRRVHVDQFLPPVQLPAQLRPGDQSDSGSEGEMEEPTVDHYQLGDLVAGLSHLEELDLVYDVKDCGMNFEWNLFLFTYRDCLSLAAAIKACHTLKIFKLTRSKVDDDKARIII | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit