missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TCTE1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112963
This item is not returnable.
View return policy
Description
TCTE1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
TCTE1 is a protein coding gene.
Specifications
| TCTE1 | |
| Polyclonal | |
| Unconjugated | |
| TCTE1 | |
| D17sil1; D6S46; Drc5; Dynein regulatory complex subunit 5; FAP155; t-complex-associated testis expressed 1; T-complex-associated testis-expressed protein 1; t-complex-associated-testis-expressed 1; Tcte1; tcte-1 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 202500, 21645 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1.5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| A6H639, Q5JU00 | |
| TCTE1 | |
| Recombinant Human Dynein regulatory complex subunit 5 protein (285-501AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction