missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCTE3 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81956AF488
This item is not returnable.
View return policy
Description
TCTE3 Polyclonal antibody specifically detects TCTE3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| TCTE3 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 6991 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunofluorescence | |
| Alexa Fluor 488 | |
| MGC142199, T-complex testis-specific protein 3, T-complex-associated testis-expressed protein 3, t-complex-associated-testis-expressed 3, tcte-3, tctex1 domain-containing protein 3, TCTEX1D3Tctex1 domain containing 3 | |
| TCTE3 antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human TCTE3. The immunogen is located within the first 50 amino acids of TCTE3. Amino Acid Squence: APVTPRKERRPSMFEKEAY | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction