missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCTN2 Antibody (OTI1C1), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74483FR
This item is not returnable.
View return policy
Description
TCTN2 Monoclonal specifically detects TCTN2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| TCTN2 | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| TCTN2 | |
| Full length recombinant protein of human TCTN2 (NP_079085) produced in HEK293T cells. | |
| 0.1 mL | |
| 79867.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI1C1 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| C12orf38, chromosome 12 open reading frame 38, FLJ12975, TECT2tectonic 2, tectonic family member 2, tectonic-2 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction