missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCTN2 Antibody (OTI1C1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-02824
This item is not returnable.
View return policy
Description
TCTN2 Monoclonal specifically detects TCTN2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| TCTN2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500, Flow Cytometry 1:100, Immunocytochemistry/Immunofluorescence 1:100 | |
| TCTN2 | |
| Full length human recombinant protein of human TCTN2 (NP_079085) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| 79867 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI1C1 | |
| Unconjugated | |
| C12orf38, chromosome 12 open reading frame 38, FLJ12975, TECT2tectonic 2, tectonic family member 2, tectonic-2 | |
| Mouse | |
| 76.7 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction