Learn More
Invitrogen™ TCTN3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587479
Description
Immunogen sequence: QTQSPPSFYR AGDPILTYFP KWSVISLLRQ PAGVGAGGLC AESNPAGFLE SKSTTCTRFF KNLASSCTLD SALNAASYYN FTVLKVPRSM TDPQNMEFQV PVILTSQANA PLLAGNTCQN VVSQVTYEIE TNGTFGIQKV SVSLGQTNLT VEPGASLQQH FILRFRAFQQ STAASLTSPR SGNPGYIVGK PLLALTDDIS YSMTLLQSQG NGSCSVKRHE VQFGVNAISG CKLRLKKADC SHLQQEIYQT LHGRPRPEYV AIFGNADPAQ KGGWTR; Positive Samples: Mouse craniofacial, Mouse lung, Mouse brain; Cellular Location: Membrane, Single-pass type I membrane protein.
TCTN3 is a member of the Tectonic protein family, a group of evolutionarily conserved secreted and transmembrane proteins that regulate the Hedgehog (Hh)-mediated patterning of the neural tube. While the expression pattern and function of TCTN3 is not known, it is 58% similar to TCTN1. TCTN1 is expressed during embryonic development in regions that participate in Hh signaling, beginning in the gastrulation stages in the ventral node. Mice expressing mutant TCTN1 die between E13.5 and E16.5 and display holoprosencephaly, a defect associated with reduced Hh signaling, indicating the role of TCTN1 as an Hh activator. At later stages in development, TCTN1 is thought to also act as a repressor on the Hh pathway in the anterior and posterior neural tube.
Specifications
| TCTN3 | |
| Polyclonal | |
| Unconjugated | |
| TCTN3 | |
| 4930521E07Rik; AI197391; C10orf61; JBTS18; OFD4; PSEC0041; RGD1305166; RP11-7D5.3; TCTN3; Tect3; tectonic 3; tectonic family member 3; tectonic-3; UNQ1881/PRO4324 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 26123, 67590 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.85 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6NUS6, Q8R2Q6 | |
| TCTN3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 200-475 of human TCTN3 (NP_0564464). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.