missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TDG Antibody (2F7), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22589B
This item is not returnable.
View return policy
Description
TDG Monoclonal antibody specifically detects TDG in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| TDG | |
| Monoclonal | |
| Biotin | |
| EC 3.2.2.29, thymine-DNA glycosylaseG/T mismatch-specific thymine DNA glycosylase | |
| Recombinant human TDG (1-410aa) purified from E. coli | |
| 0.1 mL | |
| Epigenetics, Stem Cell Markers | |
| 6996 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, Immunocytochemistry | |
| 2F7 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction