missing translation for 'onlineSavingsMsg'
Learn More

TDG Antibody (OTI3C11), FITC, Novus Biologicals™

Product Code. 18117686 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18117686 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18117686 Supplier Novus Biologicals Supplier No. NBP274484F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TDG Monoclonal specifically detects TDG in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TDG
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI3C11
Conjugate FITC
Dilution Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias EC 3.2.2.29, thymine-DNA glycosylaseG/T mismatch-specific thymine DNA glycosylase
Gene Symbols TDG
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 160-253 of human TDG (NP_003202) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Base Excision Repair, DNA Repair, Epigenetics, microRNA, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6996.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.