missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TDH Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121554
This item is not returnable.
View return policy
Description
TDH Polyclonal Antibody for Western Blot, IHC (P)
This gene appears to be an evolving pseudogene of L-threonine 3-dehydrogenase (TDH). In both prokaryotes and eukaryotes, TDH catalyzes the first of two steps in one of two L-threonine degradation pathways. However, in human, the single gene with sequence similarity to TDH is not capable of encoding a functional TDH protein; the predicted protein lacks most of the C-terminus and parts of the NAD+ binding motif when compared to other species' TDH proteins. This suggests that the human gene is therefore a pseudogene. Transcripts of this gene are found in all tissues and alternatively spliced transcripts have been described. It is not known if these transcripts are translated, or if the possible protein product provides any functional role.
Specifications
| TDH | |
| Polyclonal | |
| Unconjugated | |
| TDH | |
| L-Threonine Dehydrogenase | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 157739, 397065 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q8IZJ6, Q8MIR0 | |
| TDH | |
| Recombinant protein L-Threonine Dehydrogenase. The antigen corresponds to amino acid range 1-229 of the target protein. | |
| 20 μL | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction