missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TDO2 Antibody (OTI1D1), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74485IR
This item is not returnable.
View return policy
Description
TDO2 Monoclonal specifically detects TDO2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| TDO2 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TDO2 | |
| Full length recombinant protein of human TDO2 (NP_005642) produced in HEK293T cells. | |
| 0.1 mL | |
| 6999.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI1D1 | |
| Western Blot | |
| TDOTO, TPH2EC 1.13.11.11, TRPOtryptophanase, Tryptamin 2,3-dioxygenase, tryptophan 2,3-dioxygenase, Tryptophan oxygenase, Tryptophan pyrrolase, Tryptophanase | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction