missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TDO2 Antibody (OTI1D1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-02002
This item is not returnable.
View return policy
Description
TDO2 Monoclonal specifically detects TDO2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| TDO2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000, Immunocytochemistry/Immunofluorescence | |
| TDOTO, TPH2EC 1.13.11.11, TRPOtryptophanase, Tryptamin 2,3-dioxygenase, tryptophan 2,3-dioxygenase, Tryptophan oxygenase, Tryptophan pyrrolase, Tryptophanase | |
| Mouse | |
| 47.7 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| OTI1D1 | |
| Unconjugated | |
| P48775 | |
| TDO2 | |
| Full length human recombinant protein of human TDO2(NP_005642) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| 6999 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction