Learn More
TDP-43 Rabbit anti-Human, Unconjugated, Clone: 11N20, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech 80001-1-RR-100UL
Description
The TDP-43 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human TDP-43. This antibody recognizes human, mouse, rat antigen. The TDP-43 antibody has been validated for the following applications: FC, IHC, WB, ELISA analysis.
Specifications
| TDP-43 | |
| Monoclonal | |
| 250 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunohistochemistry, Flow Cytometry | |
| 11N20 | |
| Unconjugated | |
| 100 μL | |
| 23435 | |
| IgG |