missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TDP-43/TARDBP Antibody (DB9), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50331UV
This item is not returnable.
View return policy
Description
TDP-43/TARDBP Monoclonal specifically detects TDP-43/TARDBP in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| TDP-43/TARDBP | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TARDBP | |
| A His-tagged recombinant protein corresponding to amino acid residues 208-414 of human TDP-43/TARDBP. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| DB9 | |
| Western Blot, ELISA | |
| ALS10TAR DNA-binding protein-43, TAR DNA binding protein, TDP43, TDP-43, TDP-43TAR DNA-binding protein 43 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 23435 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction